Anti-HAO1 Mouse Monoclonal Antibody [clone: UMAB117]
Catalog # ORIGUM570082
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Hydroxyacid oxidase (glycolate oxidase) 1
- Antigen symbol:HAO1
- Clonality:Monoclonal
- Clone:UMAB117
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:40.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human HAO1 (NP_060015) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:30 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:100-200
Type: Primary
Antigen: HAO1
Clonality: Monoclonal
Clone: UMAB117
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human