Anti-L1CAM Mouse Monoclonal Antibody [clone: UMAB47]
Catalog # ORIGUM570043
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:L1CAM
- Clonality:Monoclonal
- Clone:UMAB47
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:137.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human L1CAM (NP_000416) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:30 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:100
Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: UMAB47
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human