Anti-HER2 Mouse Monoclonal Antibody [clone: UMAB36]
Catalog # ORIGUM570036
Leverantör: OriGene
Specifikationer
- Antibody type:Primary
- Antigen symbol:HER2
- Clonality:Monoclonal
- Clone:UMAB36
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:676 to 1255
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:137.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 676-1255 of human ERBB2(NP_004439) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:30 μl
- Pk:30 µl
Specifikationer
Om denna produkt
Recommended Dilutions: Western Blot:1:500; Immunochemistry1:50; Immunofluorescence:1:100
Type: Primary
Antigen: HER2
Clonality: Monoclonal
Clone: UMAB36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat