Anti-ADIPOQ Mouse Monoclonal Antibody [clone: UMAB104]
ORIGUM500072
: OriGene
- Antibody type:Primary
- Antigen name:Adiponectin
- Antigen symbol:ADIPOQ
- Clonality:Monoclonal
- Clone:UMAB104
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Amino acid number:19 to 244
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:24.9 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 19-244 of human ADIPOQ(NP_004788) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Immunochemistry: 1:100
Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: UMAB104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human