Order Entry
Sweden
ContactUsLinkComponent
Anti-SNX16 Rabbit Polyclonal Antibody
Anti-SNX16 Rabbit Polyclonal Antibody
Catalog # ORIGTA890009
Supplier:  OriGene
Anti-SNX16 Rabbit Polyclonal Antibody
Catalog # ORIGTA890009
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SNX16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    corresponding to a region within amino acids 70 to 150
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    39 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide conjugated to KLH derived from within residues 70-150 of Human SNX16.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: SNX16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human