Anti-PTRF Rabbit Polyclonal Antibody
ORIGTA320160
: OriGene
- Antibody type:Primary
- Antigen name:Polymerase I and transcript release factor
- Antigen symbol:PTRF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PTRF Antibody is supplied in PBS containing 0,02% sodium azide.
- Molecular weight:43 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Rabbit polyclonal PTRF antibody was raised against a 19 amino acid peptide near the carboxy terminus of human PTRF.
- Purification:purified by affinity chromatography via peptide column
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; IHC; Immunofluorescence
Type: Primary
Antigen: PTRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human