Anti-AKT1 Mouse Monoclonal Antibody [clone: 14E5.A2.B2.H9/E5]
Catalog # ORIGTA319564
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase B alpha
- Antigen symbol:PKBA
- Clonality:Monoclonal
- Clone:14E5.A2.B2.H9/E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:AKT
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Anti-AKT1 Antibody was produced by repeated immunizations with a synthetic peptide corresponding to internal residues of human AKT1 protein.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:1000; Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9/E5
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype:
Reactivity: Human