Anti-AHSA1 Rabbit Polyclonal Antibody
Catalog # ORIGTA319196
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Activator Of Heat Shock 90kDa Protein ATPase Homolog 1
- Antigen symbol:AHSA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Chimpanzee,Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:AHA1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human AHA1 protein.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: ELISA: 1:35000-1:185000; Western Blot: 1:500-1:3000
Type: Primary
Antigen: AHSA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human