Anti-GDF15 Rabbit Polyclonal Antibody
Catalog # ORIGTA319178
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:GDF15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This affinity purified antibody was prepared by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminal end of human NAG-1 protein. A residue of cysteine was added to facilitate coupling to KLH.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: ELISA: 1:10000; Western Blot: 2000-10000
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human