Anti-SERPINA1 Rabbit Polyclonal Antibody
Catalog # ORIGTA319137
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
- Antigen symbol:SERPINA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human Alpha-1-anti-Trypsin
- Pk:1 mg
Specifications
About this item
Recommended Dilutions: ELISA: 1:10000-1:600000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:500-1:2500
Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human