Order Entry
Sweden
ContactUsLinkComponent
Anti-CLDN6 Mouse Monoclonal Antibody [clone: 3E3]
Anti-CLDN6 Mouse Monoclonal Antibody [clone: 3E3]
Catalog # ORIGCF507011
Supplier:  OriGene
Anti-CLDN6 Mouse Monoclonal Antibody [clone: 3E3]
Catalog # ORIGCF507011
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CLDN6
  • Clonality:
    Monoclonal
  • Clone:
    3E3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    23.1 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human CLDN6(NP_067018) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: CLDN6
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human