Order Entry
Sweden
ContactUsLinkComponent
Anti-TOP2A Mouse Monoclonal Antibody [clone: 1D2]
Anti-TOP2A Mouse Monoclonal Antibody [clone: 1D2]
Catalog # ORIGCF802684
Supplier:  OriGene
Anti-TOP2A Mouse Monoclonal Antibody [clone: 1D2]
Catalog # ORIGCF802684
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TOP2A
  • Clonality:
    Monoclonal
  • Clone:
    1D2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    1100 to 1531
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    174.2 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 1100-1531 of human TOP2A(NP_001058) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human