Order Entry
Sweden
ContactUsLinkComponent
Anti-DPPA2 Mouse Monoclonal Antibody [clone: 1G10]
Anti-DPPA2 Mouse Monoclonal Antibody [clone: 1G10]
Catalog # ORIGCF800361
Supplier:  OriGene
Anti-DPPA2 Mouse Monoclonal Antibody [clone: 1G10]
Catalog # ORIGCF800361
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Developmental pluripotency associated 2
  • Antigen symbol:
    DPPA2
  • Clonality:
    Monoclonal
  • Clone:
    1G10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Dog,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    1 to 175
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    33.6 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 1-175 of human DPPA2 (NP_620170) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DPPA2
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat