Anti-THAP6 Mouse Monoclonal Antibody [clone: 2A7]
Catalog # ORIGCF800246
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:THAP6
- Clonality:Monoclonal
- Clone:2A7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:25.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human THAP6 (NP_653322) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: THAP6
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human