Anti-SCFD1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # ORIGCF504668
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:SCFD1
- Clonality:Monoclonal
- Clone:2B7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Dog,Rat
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human SCFD1(NP_057190) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500
Type: Primary
Antigen: SCFD1
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat