Anti-TMEM100 Mouse Monoclonal Antibody [clone: 2D3]
ORIGCF500499
: OriGene
- Antibody type:Primary
- Antigen symbol:TMEM100
- Clonality:Monoclonal
- Clone:2D3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:14.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full-length protein expressed in 293T cell transfected with human TMEM100 expression vector
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50
Type: Primary
Antigen: TMEM100
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human