Anti-MAPK1 Mouse Monoclonal Antibody [clone: 7E7]
Catalog # ORIGCF500486
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:MAPK1
- Clonality:Monoclonal
- Clone:7E7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Dog,Rat
- Western blot:Yes
- Epitope:full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:41.4 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human MAPK1(NP_620407) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:50-100; Immunoprecipitation: 2?g/500?l
Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 7E7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat