Order Entry
Sweden
ContactUsLinkComponent
Anti-AK1 Mouse Monoclonal Antibody [clone: 18C7]
Anti-AK1 Mouse Monoclonal Antibody [clone: 18C7]
Katalog # ORIGCF500323
Leverantör:  OriGene
Anti-AK1 Mouse Monoclonal Antibody [clone: 18C7]
Katalog # ORIGCF500323
Leverantör:  OriGene

Specifikationer

  • Antibody type:
    Primary
  • Antigen name:
    adenylate kinase 1
  • Antigen symbol:
    AK1
  • Clonality:
    Monoclonal
  • Clone:
    18C7
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    full length
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    21.6 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human AK1 (NP_000467) produced in HEK293T cell.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifikationer

Om denna produkt

Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AK1
Clonality: Monoclonal
Clone: 18C7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human