- Antibody type:Primary
- Antigen name:neurotrophic tyrosine kinase, receptor, type 2
- Antigen symbol:NTRK2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Gene ID:4915
- Antigen synonyms:GP145-TrkB|trk-B|TRKB
- UniProtKB:Q16620
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E.coli-derived human TrkB recombinant protein (Position: E66-E242). Human TrkB shares 91.5% and 89.8% amino acid (aa) sequence identity with mouse and rat TrkB, respectively.
- Purification:Immunogen affinity purified.
- Size:100 µg/vial
- Pk:100 µG
Rabbit IgG polyclonal antibody for BDNF/NT-3 growth factors receptor(NTRK2) detection. Tested with WB in Human;Mouse;Rat.
TrkB is also known as NTRK2. This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation. Mutations in this gene have been associated with obesity and mood disorders. Alternative splicing results in multiple transcript variants. Tropomyosin receptor kinase B is the high affinity catalytic receptor for several "neurotrophins", which are small protein growth factors that induce the survival and differentiation of distinct cell populations. The neurotrophins that activate TrkB are: BDNF (Brain Derived Neurotrophic Factor), neurotrophin-4 (NT-4), and neurotrophin-3 (NT-3). As such, TrkB mediates the multiple effects of these neurotrophic factors, which includes neuronal differentiation and survival.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for TrkB is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: PKM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human