Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # IMMRIR2626
Leverantör:  ImmunoReagents
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # IMMRIR2626
Leverantör:  ImmunoReagents

Specifikationer

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Storage buffer:
    10 mM Sodium phosphate, 0,15 M Sodium chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2 - 8 °C
  • Concentration:
    1,0 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2 - 8 °C
  • Immunogen:
    Mouse IgG
  • Purification:
    Affinity purified using solid phaseMouse IgG
  • Size:
    1,0 mg
  • Pk:
    1 mg

Specifikationer

Om denna produkt

ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse