Specifications
- Antikroppstyp:Sekundär
- Antikroppsnamn:Immunoglobulin G
- Antigensymbol:IgG
- Klonalitet:Polyclonal
- Konjugation:DyLight® 350
- ELISA:Yes
- Flödescytometri:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Immunutfällning:Yes
- Isotyp:IgG
- Reaktivitet:Llama
- Western blot:Yes
- Korsad adsorption:No
- Mått:Purified IgG
- Form:Lyophilized
- Storage buffer:10 mM Sodium phosphate, 0,15 M Sodium chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
- Förvaringstemperatur:2...8 °C
- Koncentration:1,0 mg/ml (E 1% at 280 nm = 13,0)
- Shipping temperature:2...8 °C
- Immunogen:Llama IgG
- Tested applications:Fluoresence ELISA, Fluorescence-Activated Cell Sorting
- Rening:Affinity purified using solid phaseLlama IgG
- Storlek:1,0 mg
- Förp.:1 mg
Specifications
About this item
The ImmunoReagents' DyLight® 350 conjugates are a derivative of AMCA (Aminomethylcoumarine acetate), which a blue fluorescent dye commonly is used as a contrasting dye for double- and triple-labeling used in a wide array of applications. The UV-excitable DyLight® 350 has spectral properties that are very similar to those displayed by AMCA, including identical excitation wavelengths and a large stokes shift. The emission wavelength of DyLight® 350 has a 10-nm blue shift over AMCA, though, that provides an advantage in multiplexing with green fluorophores. (Excitation/Emission = 353nm / 432nm Emission Color = Blue (Similar Dyes: Alexa Fluor 350, AMCA)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Llama