Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Plasminogen Goat Polyclonal Antibody
  IMMRIR2116
Anti-Plasminogen Goat Polyclonal Antibody
  IMMRIR2116

 

  • Antibody type:
    Primary
  • Antigen name:
    Plasminogen
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colourless liquid, 0,2 µm filtered
  • Antigen synonyms:
    Plasmin light chain B|DKFZp779M0222|Microplasmin|Plasmin heavy chain A|Plasmin|Angiostatin|PLG
  • Storage buffer:
    10 mM sodium phosphate, 0,15 M sodium chloride, pH 7,2
  • Storage temperature:
    2...8 °C
  • Concentration:
    >4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Human Plasminogen
  • Tested applications:
    Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
  • Purification:
    Affinity purified using solid phase Human Plasminogen
  • Size:
    1,0 mg
  • Pk:
    1 mg

 

 

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
 : Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Primary
Antigen: Plasminogen
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human