Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
  IMMRIR2011
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
  IMMRIR2011

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Lyophilised
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    10 mM sodium phosphate, 0,15 M sodium chloride, pH 7,2, 1 % (w/v) BSA, protease/IgG free
  • Storage temperature:
    2...8 °C
  • Concentration:
    1,0 mg/ml
  • Shipping temperature:
    2...8 °C
  • Immunogen:
    Mouse IgG
  • Purification:
    Affinity purified using solid phaseMouse IgG
  • Size:
    0,5 mg
  • Pk:
    0,5 mg

 

 

ImmunoReagents’ Horseradish Peroxidase (HRP) antibody conjugates are the most popular conjugates due to their high level of sensitivity, stability, and low costs. HRP conjugates are an ideal choice for use in ELISA, Immunohistochemistry (IHC), and Western blotting experiments.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
 : Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse