Order Entry
Sweden
ContactUsLinkComponent
Anti-IgE epsilon Goat Polyclonal Antibody (DyLight® 650)
Anti-IgE epsilon Goat Polyclonal Antibody (DyLight® 650)
  IMMRIR1560
undefined
Anti-IgE epsilon Goat Polyclonal Antibody (DyLight® 650)
  IMMRIR1560

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin E epsilon
  • Antigen symbol:
    IgE
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgE
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2…8 °C
  • Concentration:
    1,0 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2…8 °C
  • Immunogen:
    Human IgE, (µ) chain
  • Tested applications:
    Fluoresence ELISA, Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phase Human IgE
  • Size:
    1,0 mg
  • Pk:
    1 mg

 

 

The ImmunoReagents DyLight® 650 conjugates provide vibrant far-red fluorescence with comparable or improved performance compared to other dyes for fluorescent applications. DyLight® 650 dye conjugates have tight excitation/emission spectra that are ideal for multi-color applications. DyLight® 650 conjugated secondary antibodies are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 652 nm / 672 nm Emission Color = Far Red (Similar Dyes: Alexa Fluor 647, Cy5).

  • Origin: US

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
 : Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Goat
Isotype: IgE
Reactivity: Human