Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Chicken Polyclonal Antibody (DyLight® 550)
Anti-IgG Chicken Polyclonal Antibody (DyLight® 550)
Catalog # IMMRIR1052
Supplier:  ImmunoReagents
undefined
Anti-IgG Chicken Polyclonal Antibody (DyLight® 550)
Catalog # IMMRIR1052
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 550
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Chicken
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Lyophilised
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1% (w/v) BSA, Protease/IgG free
  • Storage temperature:
    Store freeze-dried powder at 2...8 °C
  • Concentration:
    >4,5 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified Mouse IgG, whole molecule
  • Tested applications:
    Fluoresence ELISA, Fluorescence-activated cell sorting
  • Purification:
    Affinity purified using solid phase Mouse IgG
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

The ImmunoReagents' DyLight® 550 conjugates provide vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives, including Alexa Fluor 555, TRITC and Cy3 for fluorescent applications. DyLight® 550 conjugates have tight excitation/emission spectra that are ideal for multicolor applications. DyLight® 550 conjugated secondary antibodies are used in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 562 nm / 576 nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, TRITC)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Mouse