Anti-Vitellogenin Mouse Monoclonal Antibody [clone: 6D842]
USBIV2150-58
- Antibody type:Primary
- Antigen name:Vitellogenin
- Antigen symbol:VTG
- Clonality:Monoclonal
- Clone:6D842
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 12mM sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 1:100-1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: Vitellogenin
Clonality: monoclonal
Clone: 6D842
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: