Order Entry
Sweden
ContactUsLinkComponent
Anti-VCP Rabbit Polyclonal Antibody
Anti-VCP Rabbit Polyclonal Antibody
Catalog # USBIV1035-20H
Supplier:  US Biological
Anti-VCP Rabbit Polyclonal Antibody
Catalog # USBIV1035-20H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Valosin containing protein
  • Antigen symbol:
    VCP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 1% BSA, 0,02% thimerosal, 30% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:500-1:2000

Type: Primary
Antigen: VCP (valosin containing protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse