Anti-SCGB3A1 Mouse Monoclonal Antibody [clone: 11C60]
Catalog # USBIU7001-20
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Secretoglobin Family 3A, Member 1
- Antigen symbol:SCGB3A1
- Clonality:Monoclonal
- Clone:11C60
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:21 to 104
- Storage buffer:PBS, 5% trehalose. sterile PBS.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 1-2?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: SCGB3A1 (secretoglobin, family 3A, member 1)
Clonality: monoclonal
Clone: 11C60
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human