Anti-TYK2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBIT9220-02C-HRP
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:Tyrosine Kinase 2
- Antigen symbol:TYK2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifikationer
Om denna produkt
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: TYK2 (tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human