Anti-Trypsin 3 Goat Polyclonal Antibody
Catalog # USBIT8669-70
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Trypsin 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:16 to 246
- Storage buffer:PBSwith 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: Trypsin 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse