Order Entry
Sweden
ContactUsLinkComponent
Anti-Treponema pallidum Rabbit Polyclonal Antibody (Biotin)
Anti-Treponema pallidum Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIT8300-11B
Supplier:  US Biological
Anti-Treponema pallidum Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIT8300-11B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Treponema pallidum
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~4-5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:3000

Type: Primary
Antigen: Treponema pallidum
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: