Anti-TOLLIP Rabbit Polyclonal Antibody
Catalog # USBIT8063-53
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Toll Interacting Protein
- Antigen symbol:TOLLIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 2?g/ml
Type: Primary
Antigen: TOLLIP (toll interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat