Anti-Tissue Plasminogen Activator Sheep Polyclonal Antibody
USBIT5600-05L
- Antibody type:Primary
- Antigen name:Tissue Plasminogen Activator
- Antigen symbol:TPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Reactivity:Rat,Mouse
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Mouse, Rat