Anti-TFPI2 Goat Polyclonal Antibody
USBIT5576-21A
- Antibody type:Primary
- Antigen name:Tissue Factor Pathway Inhibitor 2
- Antigen symbol:TFPI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:23 to 211
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: TFPI2 (tissue factor pathway inhibitor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse