Anti-SFTPD Mouse Monoclonal Antibody [clone: 6F188]
USBIS8402-09
- Antibody type:Primary
- Antigen name:Surfactant Protein D
- Antigen symbol:SFTPD
- Clonality:Monoclonal
- Clone:6F188
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 40% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:10000; Immunochemistry: 1:400
Type: Primary
Antigen: SFTPD (surfactant protein D)
Clonality: Monoclonal
Clone: 6F188
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human