Anti-Specific Intracellular Adhesion Molecule Grabbing Nonintegrin R1, Low Endotoxin Rat Monoclonal Antibody [clone: 10F437]
USBIS1013-69C
- Antibody type:Primary
- Antigen symbol:Specific Intracellular Adhesion Molecule Grabbing Nonintegrin R1
- Clonality:Monoclonal
- Clone:10F437
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgM
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:500 µg
- Pk:500 µG
Recommended dilution: Flow Cytometry: Immunochemistry: 1:25-1:50
Type: Primary
Antigen: Specific Intracellular Adhesion Molecule Grabbing Nonintegrin R1, Low Endotoxin
Clonality: Monoclonal
Clone: 10F437
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse