Anti-Rubella, E1 Mouse Monoclonal Antibody [clone: 10J77]
USBIR9700-05R
- Antibody type:Primary
- Antigen symbol:Rubella, E1
- Clonality:Monoclonal
- Clone:10J77
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Rubella, E1
Clonality: monoclonal
Clone: 10J77
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: