Anti-Rubella, E1 Mouse Monoclonal Antibody [clone: 9L635]
Catalog # USBIR9700-05H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Rubella, E1
- Clonality:Monoclonal
- Clone:9L635
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:0,02M Boric Acid, 5mM sodium Borate, 0,2M sodium chloride, pH 8.0+0,2, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Rubella, E1
Clonality: monoclonal
Clone: 9L635
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: