Anti-URI1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
USBIR2039-21M-AP
- Antibody type:Primary
- Antigen name:URI1, prefoldin-like chaperone
- Antigen symbol:URI1
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:250-1:500
Do not freeze!
Type: Primary
Antigen: URI1 (URI1, prefoldin-like chaperone)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human