Anti-Rho guanine nucleotide exchange factor 4 Goat Polyclonal Antibody
Catalog # USBIR1998-27B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Rho guanine nucleotide exchange factor 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris-saline, pH 7,2, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:32000; Western Blot: 2-4?g/ml
Type: Primary
Antigen: Rho guanine nucleotide exchange factor 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human