Anti-Resistin-like Molecule, beta Rabbit Polyclonal Antibody (FITC (Fluorescein))
USBIR1587-21G
- Antibody type:Primary
- Antigen symbol:Resistin-like Molecule, beta
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Amino acid number:25 to 45
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Resistin-like Molecule, beta
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse