Order Entry
Sweden
ContactUsLinkComponent
Anti-Protein Tyrosine Phosphatase, Receptor Type Z1 Chicken Polyclonal Antibody
Anti-Protein Tyrosine Phosphatase, Receptor Type Z1 Chicken Polyclonal Antibody
Catalog # USBIP9109-40Z
Supplier:  US Biological
Anti-Protein Tyrosine Phosphatase, Receptor Type Z1 Chicken Polyclonal Antibody
Catalog # USBIP9109-40Z
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Tyrosine Phosphatase, Receptor Type Z1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    430 to 733
  • Storage buffer:
    PBS, pH 7,20,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: Protein Tyrosine Phosphatase, Receptor Type Z1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human