Anti-PPM1A Mouse Monoclonal Antibody [clone: 11C45]
Catalog # USBIP9107-13E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:protein phosphatase, Mg2+/Mn2+ dependent, 1A
- Antigen symbol:PPM1A
- Clonality:Monoclonal
- Clone:11C45
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:1 to 324
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: PPM1A (protein phosphatase, Mg2+/Mn2+ dependent, 1A)
Clonality: monoclonal
Clone: 11C45
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat