Anti-Prostate Specific Gene-1 Rabbit Polyclonal Antibody
Catalog # USBIP9054-77B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Prostate Specific Gene-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Prostate Specific Gene-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human