Order Entry
Sweden
ContactUsLinkComponent
Anti-Prostaglandin E1 Rabbit Polyclonal Antibody
Anti-Prostaglandin E1 Rabbit Polyclonal Antibody
Catalog # USBIP9053-28C
Supplier:  US Biological
Anti-Prostaglandin E1 Rabbit Polyclonal Antibody
Catalog # USBIP9053-28C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Prostaglandin E1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Format:
    Serum
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Prostaglandin E1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: