Anti-Proprotein Convertase Subtilisin Kexin 9 Rabbit Polyclonal Antibody
USBIP9052-40H
- Antibody type:Primary
- Antigen name:Proprotein Convertase Subtilisin Kexin 9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: Flow Cytometry: 1:10-1:50; Immunochemistry: 1:10-1:50; Western Blot: 1:1000
Type: Primary
Antigen: Proprotein Convertase Subtilisin Kexin 9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human