Anti-Procalcitonin Mouse Monoclonal Antibody [clone: 5E757]
Catalog # USBIP9005-25G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Procalcitonin
- Clonality:Monoclonal
- Clone:5E757
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Procalcitonin
Clonality: monoclonal
Clone: 5E757
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human