Anti-Polio Virus 1,2,3 Goat Polyclonal Antibody (Biotin)
Catalog # USBIP4430-10B
Leverantör: US Biological
Specifikationer
- Antibody type:Primary
- Antigen name:Polio Virus 1,2,3
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide
- Storage temperature:Cool
- Concentration:~4 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Specifikationer
Om denna produkt
Type: Primary
Antigen: Polio Virus 1,2,3
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: