Order Entry
Sweden
ContactUsLinkComponent
Anti-PERP Rabbit Polyclonal Antibody
Anti-PERP Rabbit Polyclonal Antibody
  USBIP3353-01C
Anti-PERP Rabbit Polyclonal Antibody
  USBIP3353-01C

 

  • Antibody type:
    Primary
  • Antigen name:
    PERP, TP53 apoptosis effector
  • Antigen symbol:
    PERP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: PERP (PERP, TP53 apoptosis effector)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human