Anti-PCNA Mouse Monoclonal Antibody [clone: PC10]
Katalog # USBIP3115-16C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Proliferating cell nuclear antigen
- Antigen symbol:PCNA
- Clonality:Monoclonal
- Clone:PC10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone: PC10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human